Oakton Reagent Kit for Colorimeters, Total Chlorine and Ozone Testing; 100 Tests/Kit
Representative image only.
Oakton Item # WW-35645-66
  • High quality powder reagents for exceptional accuracy, repeatablilty, and stability
  • US-EPA approved DPD method
  • Chlorine, Total (DPD) Reagent Kit — enough for 100 tests
  • Use with the C-401, C-301, C-201, C-104 and C-105 Oakton Colorimeters
  • 3 year warranty
PRODUCT OPTIONS
₹20,200.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Test ParametersTotal chlorine and ozone
  • Number of Tests Included100
  • DescriptionReagent Kit for Colorimeters, Total Chlorine and Ozone Testing; 100 Tests/Kit
  • Warranty3 year
3 Year Warranty
Key Features
  • High quality powder reagents for exceptional accuracy, repeatablilty, and stability
  • US-EPA approved DPD method
  • Chlorine, Total (DPD) Reagent Kit — enough for 100 tests
  • Use with the C-401, C-301, C-201, C-104 and C-105 Oakton Colorimeters