Oakton pHTestr 10 Waterproof Pocket Tester 35634-10

Representative image only.
Oakton Item # WW-35634-10
Versatile, compact design works in a field or laboratory setting
  • A complete meter with key features at an economical price
  • Replaceable, double-junction electrode sensors save time and money
  • IP67-rated housing sits flat on the table and floats in water
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypeTester
  • Display typeLCD
  • Min pH (pH)-1
  • Max pH (pH)15
  • pH Resolution0.1
  • pH Accuracy±0.1 pH
  • Buffer RecognitionNIST and USA
  • CalibrationUp to 3 points
  • Min Temperature (° C)0
  • Max Temperature (° C)50
  • Min Temperature (° F)32
  • Max Temperature (° F)122
  • Temperature compensationAutomatic
  • WaterproofYes
  • Battery4 x A76 batteries (included)
  • Diameter (cm)3.81
  • Diameter (in)1 1/2
  • DescriptionWaterproof Pocket Tester 35634-10
  • Warranty1 year
1 Year WarrantyConformite EuropeenneIP 67ISO9001:2000 Certified Supplier

Key Features

  • A complete meter with key features at an economical price
  • Replaceable, double-junction electrode sensors save time and money
  • IP67-rated housing sits flat on the table and floats in water
  • 1 year warranty

More About this Item

Waterproof pHTestr 10 is great for outdoor applications such as taking pH measurements in lakes and streams. Designed to float in water, you can even attach a line to the housing for easy retrieval. Valox® sensor housing resists most chemicals.

The pHTestr 10 has a replaceable sensor for easy maintenance. Double-junction electrode sensor handles wastewater and solutions containing sulfides, heavy metals, and TRIS. Electrode sensor offers a clog-resistant Kynar® junction and a recessed, glass pH bulb to protect against breakage.

The keypad clicks to confirm operation, and allows access to a variety of functions. Calibrate up to three pH points with the auto-buffer recognition at both USA and NIST standard buffers. Additional features include a automatic temperature compensation (ATC), hold function, auto-off function, self-diagnostic error messages, and a three-segment battery-life indicator.

Meter includes: protective plastic storage case, lanyard, and batteries.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat