Oakton pH Solutions Pack; pH 4.01/7.00/10.00, Storage/Cleaning, 500 mL each

Representative image only.
Oakton Item # WW-05942-15
Save over 25% compared to ordering separately!
  • Oakton® pH Buffer Bottles have pH/temperature tables printed on the labels for accurate calibrations
  • Cleaning solution removes protein, oil, and grease contaminants—perfect for general-cleaning use too!
  • Storage solution provides proper hydration to ensure accuracy and stability
  • 2 year warranty

Call For Price
Storage & Cleaning Solutions…
Oakton® pH Electrode Storage and Cleaning Solutions
3,900.00 - ₹91,200.00INR / Pint of 1

SPECS & DESCRIPTION

Specifications & Description

  • Buffer Kit Values4.01, 7.00, and 10.01
  • Volume (mL)500
  • DescriptionpH Solutions Pack; pH 4.01/7.00/10.00, Storage/Cleaning, 500 mL each
  • Warranty2 year
Two year warranty

Key Features

  • Oakton® pH Buffer Bottles have pH/temperature tables printed on the labels for accurate calibrations
  • Cleaning solution removes protein, oil, and grease contaminants—perfect for general-cleaning use too!
  • Storage solution provides proper hydration to ensure accuracy and stability
  • 2 year warranty

More About this Item

Convenient way to order everything needed to calibrate, clean, and store pH electrodes.

• Oakton® pH Buffer Bottles have pH/temperature tables printed on the labels for accurate calibrations, and are manufactured under ISO 9000 quality guidelines and traceable to NIST standards. If unopened, shelf life is 2 years; once opened, use pH 4 and pH 7 within 90 days, pH 10 within 30 days.

• Restore proper performance and prolong probe life with Oakton® Electrode Cleaner. The cleaning solution is safe for pH and ORP probes, while being tough on junction-clogging proteins and oils.

• Use the electrode storage solution to maintain your probes, and reestablish proper hydration after cleaning or being stored dry. Designed for routine maintenance to extend pH and ORP electrode longevity—Oakton® Electrode Storage Solution is made from high quality reagents to provide optimum performance.

To obtain a Certificate of Analysis on each item within the pack, please visit each individual items' page and enter the lot number in the space provided. For example, type part number 00654-00 into the search box at the top of the page, then scroll down on the page to the CERTIFICATE OF ANALYSIS box, and enter the lot number in the space provided.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat