Oakton pH/mV Simulator, USA-NIST-Custom mV Output
Representative image only.
Oakton Item # WW-35652-00
Simulate pH buffer and mV values for quick and easy meter checks
  • pH buffer and mV value simulation allows for easy meter checks
  • Push-button operation
  • Select from USA or NIST buffers and mV output
  • 1 year warranty
₹147,000.00INR / Each
₹27,100.00 - ₹37,000.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Length (in)2.75
  • Height (in)10.375
  • Width (in)5.75
  • DescriptionpH/mV Simulator, USA-NIST-custom mV output
  • Warranty1 year
1 Year WarrantyConformite EuropeenneISO9001:2000 Certified Supplier
Key Features
  • pH buffer and mV value simulation allows for easy meter checks
  • Push-button operation
  • Select from USA or NIST buffers and mV output
  • 1 year warranty
More About this Item
Check for meter errors and ensure properly calibrated equipment with this pocket-sized pH simulator. Simple push-button operation allows for selection of USA buffers, NIST buffers, and mV output on the LED display. Additional features include 1 GΩ high impedance test, BNC connector, and low-battery indicator. Order protective rubber boot which serves as a benchtop stand separately.