Oakton pH/Ion 700 Ion 700 Benchtop Meter with Probes and NIST-Traceable Calibration

Representative image only.
Oakton Item # WW-35419-24
Designed for today's crowded laboratory benchtops!
  • Compact footprint is over 40% smaller than other benchtop meters
  • Large display permits easier readout viewing.

PRODUCT OPTIONS

₹152,000.00INR / Each

₹25,500.00 - ₹34,500.00INR / Each

Time to recalibrate? Click on the calibration service to view details, then order.
Include Electrodes Today!

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypeBenchtop
  • Display typeLCD
  • ConnectionspH: BNC; Temperature: 2.5 mm
  • Min pH (pH)-2
  • Max pH (pH)16
  • pH Resolution0.01/0.1
  • pH Accuracy±0.01 pH
  • Buffer RecognitionNIST and USA
  • CalibrationUp to 6 points
  • Min mV (mV)-2000
  • Max mV (mV)2000
  • mV Resolution0.1 or 1 mV
  • mV Accuracy±0.2 mV or 0.05%, whichever is greater
  • Min ISE0.01 ppm
  • Max ISE1999 ppm
  • ISE Resolution0.01 ppm
  • ISE Accuracy±0.5% of reading
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • Min Temperature (° F)32
  • Max Temperature (° F)212
  • Temperature compensationAutomatic or manual
  • Power (VAC)100 to 240
  • Power (Hz)50 or 60
  • Width (cm)17.4625
  • Height (cm)6.985
  • Width (in)6 7/8
  • Height (in)2 3/4
  • Probe IncludedpH and ATC probes
  • DescriptionIon 700 Benchtop Meter with Probes and NIST Calibration
3 Year Warranty

Key Features

  • Compact footprint is over 40% smaller than other benchtop meters
  • Large display permits easier readout viewing.

More About this Item

Oakton benchtop 700-series meters are designed for use in crowded laboratory settings. Compact footprint of less that 6" x 7" (16 x 18 cm) conserves valuable space, while the large, dual-display LCD provides excellent visibility—even from a distance.

Ion 700 meter measures pH, mV, relative mV, ion concentration and temperature. Automatic temperature compensation (ATC) maintains reading accuracy even with fluctuating temperatures. Models feature five-point pH calibration with automatic buffer recognition for both USA (pH 1.68, 4.01, 7.00, 10.01, 12.45) and NIST (pH 1.68, 4.01, 6.86, 9.18, 12.45) pH buffers. Meters additionally feature ion concentration calibration of up to 5 points.

Meter features include Auto-Hold, °C measurement, and easy recall of electrode slope or offset. The convenient pull-out quick reference card keeps procedures handy at all times. Use 700-series meters with any pH, ORP, or ISE electrodes with a BNC connector.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat