Oakton pH 700 Benchtop Meter with All-in-One pH Electrode and NIST-Traceable Calibration

Representative image only.
Oakton Item # WW-35419-05
Designed for today's crowded laboratory benchtops!
  • Compact footprint is over 40% smaller than other benchtop meters
  • Large display permits easier readout viewing
  • 3 year warranty

PRODUCT OPTIONS

₹87,000.00INR / Each

₹27,100.00 - ₹37,000.00INR / Each

Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypeBenchtop
  • Display typeLCD
  • ConnectionspH: BNC; Temperature: 2.5 mm
  • Min pH (pH)-2
  • Max pH (pH)16
  • pH Resolution0.01/0.1
  • pH Accuracy±0.01 pH
  • Buffer RecognitionNIST and USA
  • CalibrationUp to 6 points
  • Min mV (mV)-2000
  • Max mV (mV)2000
  • mV Resolution0.1 or 1 mV
  • mV Accuracy±0.2 mV or 0.05%, whichever is greater
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • Min Temperature (° F)32
  • Max Temperature (° F)212
  • Temperature compensationAutomatic or manual
  • Power (VAC)100 to 240
  • Power (Hz)50 or 60
  • Width (cm)17.4625
  • Height (cm)6.985
  • Width (in)6 7/8
  • Height (in)2 3/4
  • DescriptionBenchtop Meter with \"All-in-One\" pH Electrode and NIST Calibration
  • Warranty3 year
3 Year Warranty

Includes

meter, all-in-one pH electrode, electrode stand, universal 110/240 VAC power supply and adapters, and NIST calibration report.

Key Features

  • Compact footprint is over 40% smaller than other benchtop meters
  • Large display permits easier readout viewing
  • 3 year warranty

More About this Item

The easy-to-use, microprocessor-based pH 700 benchtop meter measures pH, mV, relative mV, and temperature. The large, dual-display LCD provides excellent visibility—even from a distance. Automatic temperature compensation (ATC) maintains reading accuracy even with fluctuating temperatures. Five-point pH calibration, with automatic buffer recognition for both USA (pH 1.68, 4.01, 7.00, 10.01, 12.45) and NIST (pH 1.68, 4.01, 6.86, 9.18, 12.45) pH buffers, makes for easy calibration no matter which requirement you need to meet.

Meter features HOLD, ATC, °C measurement, recall of electrode slopand offset, and electrode diagnostics. The convenient pull-out quick reference card keeps procedures handy at all times. Meter accepts any pH or ORP electrodes with a BNC connector.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat