This item is now discontinued. We recommend the following as suggested replacements: WW-35660-12

Oakton pH 600 Waterproof Meter Kit with NIST-Traceable Calibration

Representative image only.
Oakton Item # WW-35418-71
Expanded range, resolution and accuracy, increased memory, and advanced communications!
  • No more excuses for missing data!
  • Out-of-date or unperformed calibrations are now things of the past!
  • Don't waste time looking for those cables!
  • Use in dirty and wet environments
  • 3 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypePortable
  • Display typeLCD
  • Min pH (pH)-2
  • Max pH (pH)20
  • pH Resolution0.01/0.1
  • pH Accuracy±0.01 pH
  • Buffer RecognitionDIN, NIST, and USA
  • CalibrationUp to 6 points
  • Min mV (mV)-1999
  • Max mV (mV)1999
  • mV Resolution0.1 or 1 mV
  • mV Accuracy±0.2 mV or 0.05%, whichever is greater
  • Min Temperature (° C)-10
  • Max Temperature (° C)110
  • Min Temperature (° F)14
  • Max Temperature (° F)230
  • Temperature compensationAutomatic or manual
  • InterfaceRS-232
  • Memory Size500 data sets
  • WaterproofYes
  • Battery4 x AA batteries (included)
  • Width (cm)8.255
  • Height (cm)18.415
  • Width (in)3 1/4
  • Length (in)2 1/4
  • Height (in)7 1/4
  • DescriptionWaterproof Meter Kit with NIST-traceable Calibrations
  • Warranty3 year
3 Year WarrantyIP 67ISO9001:2000 Certified Supplier

Key Features

  • No more excuses for missing data!
  • Out-of-date or unperformed calibrations are now things of the past!
  • Don't waste time looking for those cables!
  • Use in dirty and wet environments
  • 3 year warranty

More About this Item

These waterproof handheld meters feature a –2.00 to 20.00 pH range with 0.01 pH resolution. Combined with accuracies up to ±0.01 pH, these meters provide a new benchmark in our pH meter line. Store up to 500 data sets in the meter‘s memory. Time-and-date stamping meets Good Laboratory Practice (GLP) standards.

The pH 600 meters offer the latest in pH calibration capabilities including up to six calibration points, a user-settable “calibration due” alarm, and up to 15 buffer options with auto-buffer recognition of USA, NIST, DIN, and PWB standards.

The infrared (IrDA) wireless technology makes PC downloading more convenient and easier. Dual powerup allows you to use batteries while out in the field or the optional AC power adapter while in the lab.

As always, our waterproof meters are IP67 rated for protection against both dust and water, even with no probes attached.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat