Oakton pH 550 Benchtop pH Meter Kit with Probe, Stand, and pH Buffers

Representative image only.
Oakton Item # WW-35419-28
Economical meter for benchtop pH measurement
  • Automatically recognizes calibration solutions
  • Compact design saves valuable benchtop space
  • Data logging allows you to store and recall up to 50 groups of data
  • IP54-rated housing is dust- and spill-proof
  • Bundled kit for convenience and savings!
  • All Oakton pH buffers are traceable to NIST standard reference materials
  • Buffer solutions help you calibrate pH instruments to ensure accuracy
  • 3 year warranty

PRODUCT OPTIONS

₹77,500.00INR / Pkg of 1

₹27,100.00 - ₹37,000.00INR / Each

Time to recalibrate? Click on the calibration service to view details, then order.
Storage & Cleaning Solutions…
Oakton® pH Electrode Storage and Cleaning Solutions
3,900.00 - ₹91,200.00INR / Pint of 1

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypeBenchtop
  • Display typeLCD
  • Min pH (pH)0
  • Max pH (pH)14
  • pH Resolution0.01/0.1
  • pH Accuracy±0.01 pH ±1 digit
  • Buffer RecognitionNIST and USA
  • CalibrationUP to 3 points
  • Min mV (mV)-1999
  • Max mV (mV)1999
  • mV Resolution1
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • Min Temperature (° F)32
  • Max Temperature (° F)212
  • Temperature Resolution0.1/1°F (0.1°C)
  • Temperature Accuracy±0.5°C ±1 digit
  • Temperature compensation Automatic or manual from 32 to 122°F (0 to 50°C)
  • Data Logging (points)50 groups of data
  • Length (cm)23.32
  • Width (cm)15.24
  • Height (cm)6.35
  • Width (in)6
  • Length (in)8
  • Height (in)2 1/2
  • DescriptionBenchtop pH Meter Kit with Probe, Stand, and pH Buffers
  • Warranty3 year
3 Year WarrantyConformite EuropeenneIP54 Rated

Includes

Plastic 3-in-1 combination pH electrode, electrode holder, pH standard buffer solutions (4.00/7.00/10.01 pH, 500 mL each), and 9 VAC power adapter.

Key Features

  • Automatically recognizes calibration solutions
  • Compact design saves valuable benchtop space
  • Data logging allows you to store and recall up to 50 groups of data
  • IP54-rated housing is dust- and spill-proof
  • Bundled kit for convenience and savings!
  • All Oakton pH buffers are traceable to NIST standard reference materials
  • Buffer solutions help you calibrate pH instruments to ensure accuracy
  • 3 year warranty

More About this Item

The PH 550 benchtop pH meter features a microprocessor chip which provides automatic temperature compensation, data storage, parameter setup, and Max/Min reading display. The meter offers a pH range from 0 to 14 and a three-point automatic calibration, with the ability to recognize standard buffer solutions (USA or NIST series). The advanced digital processing technology improves meter response time and measuring accuracy. A stable reading display icon appears when the measuring value is stable.

The 3-in-1 combination pH electrode measures pH and temperature simultaneously. The compact meter includes a detachable electrode holder and three bottles of pH buffer solution. It has a dustproof and spillproof rating of IP54. Connectors are protected by silicone seal caps.

Oakton® pH Buffer Bottles have pH/temperature tables printed on the labels for accurate calibrations
• Manufactured under ISO 9000 quality guidelines and traceable to NIST standards
• If unopened, shelf life is 2 years; once opened, use pH 4 and pH 7 within 90 days, pH 10 within 30 days
• Store buffers away from direct sunlight.
• Color coded for quick visual reference:
- pH 4.01 buffer in red
- pH 7.00 buffer in yellow
- pH 10.01 buffer in blue

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat