Oakton ORP electrode; double-junction, BNC Connector
Representative image only.
Cole-Parmer Item # WW-59001-77
Combine durability with reliability
  • Epoxy-body housing resists impact
  • Ideal for general purpose lab and field applications
  • Compatible with most brands of pH meters
  • 6 month warranty
PRODUCT OPTIONS
₹23,000.00INR / Each
Often Bought with…
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsORP
  • Electrode TypeDouble junction
  • Electrode Body MaterialEpoxy
  • Max Temperature (° C)80
  • ConnectorBNC
  • DescriptionORP electrode; double-junction, BNC Connector
  • Warranty6 month warranty
6 Month Warranty
Key Features
  • Epoxy-body housing resists impact
  • Ideal for general purpose lab and field applications
  • Compatible with most brands of pH meters
  • 6 month warranty
More About this Item
The Ag/AgCl combination reference electrodes have an impact-resistant epoxy body. Electrodes provide a fast, stable response and are ideal for prolonged readings. Gel-filled design requires virtually no maintenance. 127 mm L x 12 mm diameter with maximum 176F (80C) temperature.