Oakton General-Purpose pH Probe, Sealed, SJ, Epoxy, Direct Connect, BNC
Representative image only.
Oakton Item # WW-35804-00
Outstanding value at an economical cost!
  • Compatible with all Oakton brand pH meters
  • Ideal for general purpose measurements
  • Direct connect
  • Impact resistant
  • Single junction type
  • 1 year warranty
PRODUCT OPTIONS
₹9,100.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsGeneral-purpose measurements
  • Electrode TypeSingle junction
  • Electrode Body MaterialEpoxy
  • Reference ElectrolyteAg/AgCl
  • Min pH (pH)0
  • Max pH (pH)12
  • Min Temperature (° C)0
  • Max Temperature (° C)80
  • ConnectorBNC
  • Diameter (mm)12
  • Length (mm)110
  • DescriptionGeneral-Purpose pH Probe, Sealed/SJ/Epoxy/Direct Connect; BNC
  • Warranty1 year
1 Year Warranty
Key Features
  • Compatible with all Oakton brand pH meters
  • Ideal for general purpose measurements
  • Direct connect
  • Impact resistant
  • Single junction type
  • 1 year warranty
More About this Item
Oakton combination electrodes are available in a variety of styles to meet the needs of your application. Epoxy electrodes are impact resistant and provide excellent durability for field applications. Glass electrodes withstand aggressive chemicals. Choose junction and tip style based on the needs of your application. All electrodes feature Ag/AgCl reference cells.