Oakton Electrode Fill Solution, 3.8M KCL, 2/pack
Representative image only.
Oakton Item # WW-05992-47
Effortless refilling
  • Choose your solution: 3.8 KCI with AgCI for Ag/AgCl electrodes or 4 KCI for calomel
  • Sold in packs of two 125-mL bottles
  • 1 year warranty
PRODUCT OPTIONS
₹4,600.00INR / Pkg of 2
Often Bought with…
SPECS & DESCRIPTION
Specifications & Description
  • Volume (mL)125
  • DescriptionElectrode Fill Solution, 3.8M KCL, 2/pack
  • Warranty1 year
1 Year Warranty
Key Features
  • Choose your solution: 3.8 KCI with AgCI for Ag/AgCl electrodes or 4 KCI for calomel
  • Sold in packs of two 125-mL bottles
  • 1 year warranty
More About this Item
Choose from two types of reference fill solutions: 3.8 M KCl saturated with AgCl for AgCl/AgCl electrodes, or 4 M KCl for calomel (Hg/Hg22) electrodes. Both solutions come in a pack of two 125-mL — enough to fill approximately 25 electrodes.