Oakton ECCONCTP0500 COND 500 Conductivity Transmitter

Representative image only.
Oakton Item # WW-19505-20
Economical and compact—yet ideal for tough applications
  • Weatherproof IP65-rated housing—suitable for harsh environments
  • Displays conductivity and temperature
  • Two wire design with input-to-output isolation

₹85,200.00INR / Each

₹18,700.00INR / Each

for an additional
₹18,700.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • Control TypeOn/Off
  • Display TypeLCD
  • Temperature CompensationAutomatic or Manual
  • Output Current4-20mA
  • Battery12 to 24 VDC
  • Width (cm)9.53
  • Height (cm)9.53
  • Width (in)3 3/4
  • Height (in)3 3/4
  • Depth (in)2
  • DescriptionCOND 500 Conductivity Transmitter

Key Features

  • Weatherproof IP65-rated housing—suitable for harsh environments
  • Displays conductivity and temperature
  • Two wire design with input-to-output isolation

More About this Item

Simple and reliable transmitters are ideal for connecting to PLC for advanced control systems. Or, connect to a recorder for a low cost documentation system. Scaleable 4-20 mA output with automatic temperature compensation.

Install in series with each transmitter isolated so there is no disruption if one should fail. Housing contains mounting holes for easy installation on plates, poles, or walls. Knockouts with rubber septums allow wires to be connected from any direction, but keep dust and moisture out. Order conductivity cells separately from accessories.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat