Oakton Conductivity Calibration Pouches, 10 µS; 20/Pk
Representative image only.
Oakton Item # WW-35653-09
Sealed for freshness and easy travel
  • Tear off a corner, or the entire top—opens wide enough for all probe types
  • Notched corners for easy opening from either side
  • 1 year warranty
PRODUCT OPTIONS
₹7,800.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Conductivity Value (µS)10
  • Resistivity Value (MΩ)0.1
  • Volume (mL)20
  • Volume (oz)0.7
  • Volume20 pouches
  • Conductivity-to-TDS Calibration Values (ppm 442)7
  • Conductivity-to-TDS Calibration Values (ppm KCl)4.8
  • Conductivity-to-TDS Calibration Values (ppm NaCl)4.7
  • Descriptionconductivity calibration pouches, 10 µS, box of 20 pouches
  • Warranty1 year
1 Year Warranty
Key Features
  • Tear off a corner, or the entire top—opens wide enough for all probe types
  • Notched corners for easy opening from either side
  • 1 year warranty
More About this Item
Single-use 20-mL pouches are sealed to ensure freshness—take these convenient standard solutions anywhere! Don't worry about transferring solution into another container—calibrate directly from the pouch.
A µS-to-ppm conversion table is printed on each pack for quick reference.
Certificate of Analysis
Enter lot number
or click to view a Sample C of A