Oakton by Cole-Parmer® pH electrode, Gel-Filled
Representative image only.
Oakton Item # WW-05993-25
PTFE reference-junction offer extended electrode life
  • Impact resistant—epoxy-body housing for added strength
  • Ideal for your general-purpose applications in the lab or field
  • Compatible with most brands of pH meter
  • 1 year warranty
PRODUCT OPTIONS
₹16,200.00INR / Each
Often Bought with…
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsGeneral use-low cost
  • Electrode TypeSingle junction
  • Electrode Body MaterialEpoxy
  • Reference ElectrolyteAg/AgCl
  • Max Temperature (° C)100
  • ConnectorBNC
  • DescriptionpH electrode, Gel-Filled
  • Warranty1 year
1 Year Warranty
Key Features
  • Impact resistant—epoxy-body housing for added strength
  • Ideal for your general-purpose applications in the lab or field
  • Compatible with most brands of pH meter
  • 1 year warranty
More About this Item
These quality electrodes have a sealed gel reference unless otherwise noted.