Oakton by Cole-Parmer® ORP electrode, in-line, single-junction, 10-ft cable, BNC
Representative image only.
Oakton Item # WW-05993-62
Integral, one piece electrode and housing for installation convenience
  • Choose from single-junchtion or double-junction designs
  • Available with BNC or tinned ends
  • 1 year warranty
PRODUCT OPTIONS
₹23,200.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Electrode TypeSingle junction
  • Electrode Body MaterialPVDF
  • Max Temperature (° C)80
  • ConnectorBNC
  • DescriptionORP electrode, in-line, single-junction, 10-ft cable, BNC
  • Warranty1 year
1 Year WarrantyKeep a Spare
Key Features
  • Choose from single-junchtion or double-junction designs
  • Available with BNC or tinned ends
  • 1 year warranty
More About this Item
In-line electrodes operate from 23 to 212°>F (-5 to 100°>C) and have a chemical-resistant PVDF housing with a 1/2'' NPT(M) thread. Housing measures 5'' x 15/16'' diameter. Maximum pressure 100 psi. Electrode extends 1'' beyond housing.