Oakton ATC probe for pH 600, pH 610, pH 620, PC 650, PD 650, PCD 650 meters, 3-m
Representative image only.
Oakton Item # WW-35418-07
Provides ATC for most current Oakton pH meters
  • Ensure optimal pH and ion measurement with ATC probes
  • Longer 3-m cable
  • 6 month warranty
PRODUCT OPTIONS
₹13,100.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionATC probe for pH 600, pH 610, pH 620, PC 650, PD 650, PCD 650 meters, 3-m
  • Warranty6 month
6 Month Warranty
Key Features
  • Ensure optimal pH and ion measurement with ATC probes
  • Longer 3-m cable
  • 6 month warranty
More About this Item
Corrosion-resistant stainless steel 30 KΩ thermistor features durable polymer handle and 3.3 ft (1-m) cable. Probes measure 88-mm L x 3-mm dia; handle adds another 67 mm to the total length. Temperature range is 0 to 100°C.