Oakton All-in-One pH/ATC Electrode, Epoxy, Sealed, Single-Junction, 12 mm OD; 3 ft Sub-Mini/BNC

Representative image only.
Oakton Item # WW-35811-71
Update, upgrade, or replace electrodes to get the best out of Oakton® meters
  • Choose from pH or ORP electrodes
  • 6 month warranty

PRODUCT OPTIONS

₹9,700.00INR / Each
Oakton Electrode Storage Solution; 500 mL
₹3,900.00INR / Pint of 1
Don't Forget Buffer Solutions!
Oakton® pH Buffer Solutions
1,300.00 - ₹21,000.00INR / Pint of 1

SPECS & DESCRIPTION

Specifications & Description

  • MaterialEpoxy
  • Compatible WithOakton pH meters accepting 2.5 mm audio plug for temperature
  • Length (mm)115
  • Diameter (mm)12
  • DescriptionAll-in-one pH/ATC Probe, Sealed/SJ/Epoxy, 3-ft Sub-mini/BNC; 12 mm Dia
  • Warranty6 month
6 Month Warranty

Key Features

  • Choose from pH or ORP electrodes
  • 6 month warranty

More About this Item

Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat