nVent Hoffman N170246G010 Enclosure 2000 BTU 460V 50 60 HZ

Representative image only.
nVent Hoffman Mfr # N170246G010 – Item # WW-10500-20
  • 1 year warranty

PRODUCT OPTIONS

₹929,000.00INR / Each

SPECS & DESCRIPTION

Specifications & Description

  • MaterialSteel
  • Outside Width (in)12
  • Outside Height (in)17.625
  • Outside Depth (in)8.375
  • DescriptionEnclosure 2000 BTU 460V 50 60 HZ
  • Warranty1 Year
Conformite Europeenne

Key Features

  • 1 year warranty

More About this Item

Parts and accessories to keep your Pentair enclosure working reliably.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat