NPS Sustayn by Spilfyter Recycled Universal Black Heavyweight Drum Top, 22" diameter, 25/Case
Representative image only.
NPS Mfr # G-76 – Item # WW-86423-06
Sorbent material is 100% recycled polypropylene
₹21,200.00INR / Case of 25
SPECS & DESCRIPTION
Specifications & Description
  • Absorption11 gal. (42 L)
  • OD (in)22
  • ColorBlack
  • DescriptionBlack, Heavyweight, Drum Top, 25/Case
More About this Item
Go green and support your environmental initiatives with these 100% recycled polypropylene drum spill pads. Packaging material is also recycled and printing uses environmentally-friendly water-based inks. Universal sorbents are ideal for environments where there are many different liquids including cutting oils, lubricants, and coolants. Works with water-based liquids, oil-based liquids, and more.