NPS Spilfyter Kolor-Safe Acid Neutralizer, 1 quart
Representative image only.
NPS Mfr # 410001 – Item # WW-86423-10
Three-color change lets you know when spill is completely neutralized
PRODUCT OPTIONS
₹3,700.00INR / Quart of 1
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionAcid Neutralizer, 1 quart
More About this Item
Protect your workers and facility from acid spills by quickly and efficiently neutralizing them. Simply apply the neutralizer until the spill goes through the three-color change. Acid neutralizer changes from purple to yellow to a reddish color to indicate total neutralization.