NPS Spilfyter Grab & Go Oil-Only Spill Kit, Absorbs 13 gallons, 1/Bag
Representative image only.
NPS Mfr # 311002 – Item # WW-86424-10
Kit includes everything you need for quick spill clean up, in a handy zipper bag
SPECS & DESCRIPTION
Specifications & Description
  • Absorption13 gal. (50 L)
  • DescriptionOil-Only Spill Kit, Absorbs 13 gallons, 1/Bag
Includes
Kit includes three 3" x 10' socks, three 18" x 18" pillows, fifteen 16" x 18" pads, one pair of nitrile gloves, one pair of goggles, one disposal bag with twist tie, and one clear zipper bag.
More About this Item
Oil-Only sorbents absorb all oil-based liquids and repel water. Ideal for use with lubricants, fuels, mineral oils, and vegetable oils.