NPS Spilfyter Chemical Classifier Strips, 10/Pack
Representative image only.
NPS Mfr # 570010 – Item # WW-86424-15
Classify unknown spills in 30 seconds
PRODUCT OPTIONS
₹23,500.00INR / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionChemical Classifier Strips, 10/Pack
More About this Item
Convenient strips change color to identify risk type and classify spill for proper treatment. Compare color indicator to laminated, reusable reference chart to classify unknown liquid. Chemical classifier kit tests for acid or base, iodine, oxidizers, chlorine, fluoride, bromine and petroleum/-organic products. Wastewater classifier kit tests for acid/base, petroleum/organic solvent, hydrogen sulfide, nitrite, nitrate, and fluoride risks.