Novasina eVC-18 Water Activity Sensor Filter for Chemicals
Representative image only.
Novasina Mfr # 260 1662 – Item # WW-37910-42
  • Protect your sensor from acids, chlorine, formaldehyde, and sulfur dioxide
SPECS & DESCRIPTION
Specifications & Description
  • MaterialeVC-21
  • Compatible WithLabMaster neo
  • DescriptionTest Samples Filter for Chemicals
Key Features
  • Protect your sensor from acids, chlorine, formaldehyde, and sulfur dioxide
More About this Item
Filters and dishes for the LabMaster neo Water Activity Meter
Cole-Parmer cannot sell this product in China, Japan, Germany, France, Italy, Netherlands, Belgium, Great Britain, Spain, India, South Korea, Vietnam, Taiwan, Colombia or Brazil