North by Honeywell SSG/10 Gloves, medium, pack of 10 pair
Representative image only.
North by Honeywell Mfr # SSG/10 – Item # WW-86358-01
Use alone or as a chemically resistant liner within a rubberized glove
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNorfoil®
  • SizeMedium
  • ColorGray
  • SterileNo
  • DescriptionGloves, medium, pack of 10 pair
Conformite Europeenne
More About this Item
These flexible lightweight gloves are made of a five-layer polyethylene and ethylene vinyl alcohol copolymerconstruction that resists permeation by a wide range of aromatic, chlorinated, aliphatic solvents, ketones, and esters. Ideal for lab or industrial use. Ambidextrous.