Cole-Parmer Essentials Nickel-Plated Steel Crucible Tongs, 20" L; 1/Pk
Representative image only.
Cole-Parmer Essentials Item # WW-06443-50
₹17,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Overall Length (in)20
  • Overall Length (cm)50.8
  • MaterialNickel-plated Steel
  • SterileNo
  • DescriptionNickel-Plated Steel Crucible Tongs, 20\" L; 1/Pk
More About this Item
These nickel-plated steel tongs have positive-grip corrugated tips for safely handling dishes in muffle furnaces. Use with crucibles and dishes 1-1/2" (3.8 cm) in diameter or wider.