Nichrome V inoculating loop with handle, calibrated, 10 µL contained volume
Representative image only.
Nichrome Mfr # 3740 – Item # WW-14203-26
  • 1 month warranty
PRODUCT OPTIONS
₹5,600.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Loop Length (mm)44.5
  • Handle length (mm)152
  • Loop Length (in)1 3/4
  • Handle length (in)6
  • B and S Wire Gauge26
  • Contained Volume (µL)10
  • Handle TypeInsulated
  • DescriptionV inoculating loop with handle, calibrated, 10 µL contained volume
  • Warranty1 month
Key Features
  • 1 month warranty
More About this Item
Each loop is calibrated to APHA standards. Loops are made of 26-gauge wire. Order insulated loop and needle holder for individual loop separately.