Newport LIP275-1222-01 Balance Isolation Platform, 12" W x 22" D
Representative image only.
Newport Mfr # LIP275-1222-01 – Item # WW-11500-03
Reduce the transmission of vibrations to balances and other sensitive equipment
  • Portable and lightweight–locate your balance on any countertop or table
  • Maintains balance accuracy even in the presence of operating equipment
  • Balances to hold calibration longer and load cells are protected from damage
  • 5 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Platform Length (in)22
  • Platform Width (in)12
  • DescriptionBalance Isolation Platform, 12\" W x 22\" D
  • Warranty5 years
5 Year Warranty
Key Features
  • Portable and lightweight–locate your balance on any countertop or table
  • Maintains balance accuracy even in the presence of operating equipment
  • Balances to hold calibration longer and load cells are protected from damage
  • 5 year warranty
More About this Item
Lightweight and easy to set up, the platform isolates balances and other sensitive instruments and equipment from vibrations and mechanical shock—ideal for balances, petri dishes, and shakers.
Sturdy, gray PVC platform features a non-stick neoprene surface supported by three or four vibration isolation bearings. These advanced isolation bearings protect balances and other sensitive equipment from ambient vibrations and mechanical shocks.