Neptune 000566 Folding impeller, 3-3/4" dia (max) x 5/8" bore dia
Representative image only.
Neptune Mfr # 000566 – Item # WW-50509-20
₹41,500.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Material316 Stainless Steel
  • Impeller Size (in)3.75
  • Impeller Size (mm)95.25
  • DescriptionFolding impeller, 3-3/4\" dia (max) x 5/8\" bore dia
More About this Item
Folding impeller fits perfectly into bung openings and narrow ports—folded diameter is 1-5/8". Impeller opens fully during mixing.