Neogen COLI9858B Colitag™ MPNplate™ with Sealing Film; 100/Pk
Representative image only.
Mfr # COLI9858B – Item # WW-53200-26
A modern and simple solution for enumerating microorganisms in a liquid sample
  • Makes result reading and MPN determination simple and direct
  • Tests offer a high degree of sensitivity
  • Procedures can be completed in under a minute per sample
SPECS & DESCRIPTION
Specifications & Description
  • TestMicroorganisms
  • Tests Per Kit100
  • DescriptionColitag™ MPNplate™ with Sealing Film
Key Features
  • Makes result reading and MPN determination simple and direct
  • Tests offer a high degree of sensitivity
  • Procedures can be completed in under a minute per sample
More About this Item
The Colitag MPN plate offers a fresh approach to the traditional 15-tube serial dilution method and the statistical model of the most probable number (MPN) index. Replacing the time and labor intensive 15-tube dilution method, the device has five 10-mL wells, five 1-mL wells and five 0.1-mL wells. A 16th well for collecting the remaining sample allows a full 100-mL sample to be analyzed. A mixture of Colitag reagent and sample is poured directly into the wells, filling all 15 wells first and then the overflow well. The device is then sealed with an adhesive film and set up for incubation.
The procedure requires an incubator, long-wave UV lamp, sealing film, and sealing tool.