Neogen COLI9850 Colitag™ Test Kit, P/A 100-mL Format; 20/Pk
Representative image only.
Mfr # COLI9850 – Item # WW-53200-16
Quickly and confidently analyze drinking water
  • Colitag is a presence-absence and MPN enzyme substrate test
  • Detects as few as 1 MPN of total coliform or E. coli in a 100-mL water sample
  • US EPA approved; depended on in over 24 countries
  • Maximizes flexibility; use one test instead of three
PRODUCT OPTIONS
₹16,900.00INR / Pkg of 20
SPECS & DESCRIPTION
Specifications & Description
Key Features
  • Colitag is a presence-absence and MPN enzyme substrate test
  • Detects as few as 1 MPN of total coliform or E. coli in a 100-mL water sample
  • US EPA approved; depended on in over 24 countries
  • Maximizes flexibility; use one test instead of three
More About this Item
Enhance your laboratory efficiency—and ensure water purity—with Colitag, the revolutionary 16-48 hour water test. To use, simply add Colitag to a 100-mL sample, incubate, visually check for coliforms and illuminate with a UV lamp, as bright blue fluorescence indicates a positive E. coli sample.