Needle-Point/Pic Sampler Probe, chrome-plated brass, 11" insertion depth
Representative image only.
Item # WW-33687-30
Ideal for sampling powders and crystals
  • Materials of construction: chromium-plated brass probe, aluminum handle
  • Sterilize: yes
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Length (in)11
  • DescriptionNeedle-Point/Pic Sampler Probe, chrome-plated brass, 11\" insertion depth
  • Warranty1 year
1 Year Warranty
Key Features
  • Materials of construction: chromium-plated brass probe, aluminum handle
  • Sterilize: yes
  • 1 year warranty
More About this Item
The needle-point sampler is designed to puncture paper or polyethylene bags with minimum damage. To use, puncture bag and insert the probe. The probe enters a maximum of 11" (28 cm) and is extracted with the sample stored inside the hollow handle. To release, tip the Pic sampler backward. A bag can be positioned over the handle for sample collection. Seal hole with adhesive tape or appropriate label.