Myron L aboratory Cup-Fill pH/TDS Tester; 2 to 12 pH and 0 to 5000 ppm
Representative image only.
Myron L Mfr # T6/PH – Item # WW-19401-01
Unique design offers rugged reliability
  • Measurement range is user-selectable
  • Chip/crack-resistant cell cup
  • Replaceable pH sensor on T6-PH model
  • Rugged conductivity electrodes
  • Automatic temperature compensation for both pH and conductivity
  • Shock-resistant readout
  • Simple-to-read analog display
  • 2 year warranty
₹137,000.00INR / Each
₹27,100.00 - ₹37,000.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
Include Buffers Today!
Oakton® pH Buffer Solution Packs
4,300.00 - ₹44,700.00INR / Pkg of 3
SPECS & DESCRIPTION
Specifications & Description
  • Meter TypePortable
  • Temperature compensationAutomatic
  • Descriptionaboratory Cup-Fill pH/TDS Tester; 2 to 12 pH and 0 to 5000 ppm
  • Warranty2 year
Two year warranty
Key Features
  • Measurement range is user-selectable
  • Chip/crack-resistant cell cup
  • Replaceable pH sensor on T6-PH model
  • Rugged conductivity electrodes
  • Automatic temperature compensation for both pH and conductivity
  • Shock-resistant readout
  • Simple-to-read analog display
  • 2 year warranty
More About this Item
These meters are compact, multi-range instruments which operate on the principle of electrical conductivity and contain a built-in cell cup to quickly determine the conductivity of almost any solution. Includes a dial calibrated in micromhos (μmhos) equivalent to the metric microsiemens (μS). These meters are completely self-contained. The built-in cell is automatically temperature-compensated. They are powered by two 9 volt batteries, good for at least 2000 tests, or one-year shelf life.