Mucksucker D8653259 drum and tank sampler, 3.5' long
Representative image only.
Mfr # D8653259 – Item # WW-99200-20
No leachable fillers or plasticizers
  • No glass parts to break for added safety and protection Easily identify layers through translucent material Easy to clean
  • 6 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Descriptiondrum and tank sampler, 3.5' long
  • Warranty6 month
6 Month Warranty
Key Features
  • No glass parts to break for added safety and protection Easily identify layers through translucent material Easy to clean
  • 6 month warranty
More About this Item
High-suction syringe action is ideal for viscous fluids. Efficiently draws samples that are close to the bottom of the drum or tank. Comes with both a push/seal bottom and a coring bottom. Coring bottom features a wide mouth for handling sludge samples.