Monarch 6280-028 Replacement Xenon Lamps for Stroboscopes, L-1905; 2/Pk
Representative image only.
Monarch Mfr # 6280-028 – Item # WW-53510-05
Removable, rechargeable battery pack allows unlimited measuring power
  • Compact, lightweight housing allows for convenient one-handed operation
  • Four-quadrant tuner makes stop action and slow motion inspections easy
PRODUCT OPTIONS
₹18,800.00INR / Pkg of 2
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionReplacement Xenon Lamps for Stroboscopes, L-1905; 2/Pk
Key Features
  • Compact, lightweight housing allows for convenient one-handed operation
  • Four-quadrant tuner makes stop action and slow motion inspections easy
More About this Item
The PalmStrobe x portable stroboscope is the industrial standard for high intensity, multifunction xenon stroboscopes. Memory holds eight commonly used flash patterns and the last flash rate used - no need to recalculate flash rates each time. Memory is retained even when the battery is recharging - recharge battery in or out of strobe.
The 6-digit, backlit LCD display is easy to read in both well-lit or dim locations. Additional features include rapid x2 and ÷2 of current flash rate, TTL-compatible input/output, and low-battery indicator.