Mettler Toledo Routine Pt1000 pH Electrode; MultiPin™
Representative image only.
Mettler Toledo Mfr # 51343056 – Item # WW-35900-24
Expertly-engineered probes to cover a wide range of applications
  • Cable not included⌡ARGENTHAL™ reference system⌡HA glass membrane for high pH and temperature applications
PRODUCT OPTIONS
₹112,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsHigh pH
  • Electrode Body MaterialGlass
  • Reference JunctionCeramic
  • Reference Electrolyte3M KCl
  • Sensing Glass TypeHA - High alkalifor high pH and high temp applications
  • Min pH (pH)0
  • Max pH (pH)14
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • Temperature CompensationYes
  • ATC Element1000Ω Pt RTD
  • ConnectorMultipin™
  • Diameter (mm)12
  • Length (mm)120
  • DescriptionpH Electrode; MultiPin™
Key Features
  • Cable not included⌡ARGENTHAL™ reference system⌡HA glass membrane for high pH and temperature applications
More About this Item
These ISM® - Intelligent Sensor Management electrodes models store calibration data and automatically transfer the data to your meter. Choose open junction versions for dirty and complex samples. Cables not included with all electrodes. For additional versatility—select the one that connects to your meter.
Select the appropriate sensor cable for your application, meter, and ATC connection—BNC, waterproof BNC, DIN, and Lemo OO cables available with banana or RCA (cinch) connectors. High alkali glass membrane provides low-alkali error—ideal for high pH and high temperature applications. ARGENTHAL™ reference system has a silver ion trap, for silver-ion-free electrolyte. No clogging junction from sulfides or protein binding—perfect for biological samples and TRIS buffers.