Mettler Toledo Refracto 30GS Portable Handheld Refractometer
Representative image only.
Mettler Toledo Mfr # 51324660 – Item # WW-33333-38
Reliable results within seconds
  • Ensure accuracy with the built-in temperature compensation
  • Quickly view Brix or refractive index measurements with the click of a button
  • Easily transfer important data to a PC or printer for documentation
  • Ensure accuracy with the built-in temperature compensation
  • Quickly view Brix or refractive index measurements with the click of a button
  • Easily transfer important data to a PC or printer for documentation
  • 1 year warranty
PRODUCT OPTIONS
₹1,256,000.00INR / Each
₹20,400.00 - ₹27,100.00INR / Each
for an additional
₹20,400.00INR / Each
for an additional
₹27,100.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
Calibration Standard Solution Sets
Refractive Index Standard Solutions
127,000.00 - ₹383,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Min Brix Range (%)0
  • Max Brix Range (%)85
  • Min Refractive Index1.32
  • Max Refractive Index1.65
  • Max Temperature (° F)104
  • Min Temperature (° F)50
  • Max Temperature (° C)40
  • Min Temperature (° C)10
  • Resolution0.1% Brix, 0.0001 RI
  • Accuracy±0.2% Brix, ±0.0005 RI
  • Temperature Resolution0.1C
  • Max Temperature Compensation (° F)104
  • Min Temperature Compensation (° F)50
  • Max Temperature Compensation (° C)40
  • Min Temperature Compensation (° C)10
  • Display TypeLC-Display
  • Battery Type2 AAA (included)
  • Range0 to 85% Brix, 1.32 to 1.65 RI
  • DescriptionMettler Toledo Refracto 30GS Portable Handheld Refractometer
  • Warranty1 year
1 Year WarrantyISO 9001:2008 Certified Supplier
Includes
carrying case, 2 AAA batteries, 5 cleaning tissues, 2 pipettes, 2 vials with screw cap, 1 CD-ROM
Key Features
  • Ensure accuracy with the built-in temperature compensation
  • Quickly view Brix or refractive index measurements with the click of a button
  • Easily transfer important data to a PC or printer for documentation
  • Ensure accuracy with the built-in temperature compensation
  • Quickly view Brix or refractive index measurements with the click of a button
  • Easily transfer important data to a PC or printer for documentation
  • 1 year warranty
More About this Item
The Mettler Toledo 30PX and 30GS portable refractometers are suited for determining the refractive index of liquids, providing temperature-compensated results. These are handheld and benchtop refractometers in one, and the cell can be immersed directly in the sample or a sample added to the cell for increased flexibility. Both options allow you to save 1100 results – including information such as sample and instrument identification, measurement unit, temperature correction coefficient, date, and time. Both units feature a backlit LCD for easy-to-read measurements, and can be interfaced to a PC or printer for exporting data.
The 30GS model features a sapphire prism held by a glass-plated ring for a broad measurement range.