Mettler Toledo EUP Automated Performance Verification Module, European Pharmacopeia
Representative image only.
Mettler Toledo Mfr # 30254732 – Item # WW-83700-92
Enhance performance of your spectrophotometer
  • Suitable for use with the METTLER TOLEDO Excellence UV/Visible Scanning Spectrophotometers
  • CertiRef™ automated performance verification module with EUP certified standards
  • 3 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithMettler Toledo UV5
    Excellence Spectrophotometers
  • DescriptionEUP Automated Performance Verification Module, European Pharmacopeia
  • Warranty3 month
3 Month Warranty
Key Features
  • Suitable for use with the METTLER TOLEDO Excellence UV/Visible Scanning Spectrophotometers
  • CertiRef™ automated performance verification module with EUP certified standards
  • 3 month warranty
More About this Item
Accessories are compatible with the METTLER TOLEDO Excellence UV/Visible Scanning Spectrophotometers, models 83700-00 through -03.