Mettler Toledo 8-Place Automated Sample Changer
Representative image only.
Mettler Toledo Mfr # 30236313 – Item # WW-83700-91
Enhance performance of your spectrophotometer
  • Suitable for use with the METTLER TOLEDO Excellence UV/Visible Scanning Spectrophotometers
  • Cuvette changer automatically moves up to eight cuvettes into position for measurements
  • Thermostable
  • 3 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithMettler Toledo UV5
    Excellence Spectrophotometers
  • Description8-Place Automated Sample Changer
  • Warranty3 month
3 Month Warranty
Key Features
  • Suitable for use with the METTLER TOLEDO Excellence UV/Visible Scanning Spectrophotometers
  • Cuvette changer automatically moves up to eight cuvettes into position for measurements
  • Thermostable
  • 3 month warranty
More About this Item
Accessories are compatible with the METTLER TOLEDO Excellence UV/Visible Scanning Spectrophotometers, models 83700-00 through -03.