Mettler Toledo 30300915 ErgoSens USB Optical Sensor for XSR Balance
Representative image only.
Mettler Toledo Mfr # 30300915 – Item # WW-11335-93
Optimize performance of your Mettler Toledo balances
  • Ideal for use with XSR analytical balances
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionErgoSens USB for XPR Balance
Key Features
  • Ideal for use with XSR analytical balances
More About this Item
Perform hands-free operations such as taring, zeroing or printing with these optical sensors.
This optical infrared sensor easily connects to your Mettler Toledo XRS analytical balance via USB cable to provide hands-free operation. Avoid the risk of cross-contamination when handling sticky or toxic samples.