Meta 17020-011-XL Fluid-Resistant Men's Lab Coat, White, 40" L, X-Large
Representative image only.
Meta Mfr # 17020-011-XL – Item # WW-86468-06
Liquid quickly beads and rolls off fabric – lab coat stays dry and stain-free!
SPECS & DESCRIPTION
Specifications & Description
  • Length (in)40
  • Length (cm)101.6
  • SizeX-Large
  • MaterialPolyester / Cotton
  • ColorWhite
  • DescriptionFluid-Resistant Men's Lab Coat, White, 40\" L, X-Large
More About this Item
Lab coats are constructed of 65% polyester/35% cotton fabric and treated with NANO-CARE™ protection at a molecular level. Molecular structures, known as nano-whiskers, are attached to the tiny fabric fibers - permanently altering the performance of the fabric. This allows the fabric to repel liquids and stains without altering the look, feel, or comfort of the lab coat. Ideal for biotech, pharmaceutical, and general laboratory use.
Coats feature a five-button front closure with three exterior pockets and one interior (lower right) pocket to maximize functionality. One piece back includes a two button swing belt for added style and a better fit. Unisex.