Meriam 924BA-1 Red Fluid, 2.95SP.Gr Manometer Fluid 1LB.
Representative image only.
Meriam Mfr # 924BA-1 – Item # WW-68460-06
Accurate and dependable, as well as highly versatile
  • Unit scale extends the full width of the instrument body and is graduated on each side of the indicating column
  • Scale has a white, nonglare background with black graduations and figures
  • Table- or wall-mount style
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionRed Fluid, 2.95SP.Gr Manometer Fluid 1LB.
  • Warranty1 year
1 Year Warranty
Key Features
  • Unit scale extends the full width of the instrument body and is graduated on each side of the indicating column
  • Scale has a white, nonglare background with black graduations and figures
  • Table- or wall-mount style
  • 1 year warranty
More About this Item
Well-type manometers provide fast, direct readings of pressure, vacuum, differential, flow and liquid levels. No adding of distances above and below a zero line. All scales are graduated
to correct for the drop in well level and are full width of the instrument.
Ranges: 6, 12, 20, 30, 40, and 60 inches
Note: Must be used with glass tube manometers only. Many plastics are incompatible with this fluid.