Meiji Techno Trinocular Stereozoom Microscope System; 7x-45x; boom stand
Representative image only.
Meiji Techno Mfr # EMZ-5TR+S4100+FX+MA502 – Item # WW-48402-02
Convenient, preassembled microscope systems provide superb quality and easy ordering
  • System 48402-02 includes:
  • Stereozoom body with photo tube 48402-25
  • Boom stand and holder 48404-40
  • 10x eyepieces 48402-58
  • Limited Lifetime Warranty
SPECS & DESCRIPTION
Specifications & Description
  • Magnification7x to 45x
  • Maximum Magnification45x
  • Head ConfigurationTrinocular
  • Objectives0.7x to 4.5x
  • Eyepiece Diameter (mm)34
  • Eyepiece Magnification10x
  • EyepiecesWidefield
  • Stand TypeBoom stand
  • DescriptionTrinocular Stereozoom Microscope System; 7x-45x; boom stand
  • WarrantyLTD Life Time
Conformite EuropeenneLimited Lifetime Warranty
Key Features
  • System 48402-02 includes:
  • Stereozoom body with photo tube 48402-25
  • Boom stand and holder 48404-40
  • 10x eyepieces 48402-58
  • Limited Lifetime Warranty
More About this Item
For your added convenience, we also offer preassembled microscope systems that include a microscope body, stand, and eyepieces. All eyepieces have diameters of 34 mm OD x 25 mm ID.