Meiji Techno MX4200H Economical Compound Microscope; 4x/10x/40x/1000x; 110-240 V
Representative image only.
Meiji Techno Mfr # MT4200H – Item # WW-48400-02
Infinity-corrected optical system design for high-image quality, contrast, and flatness of field
  • Smooth-operating, ball-bearing quintuple nosepiece provides effortless objective changes
  • Wide ceramic-coated stage for smooth exchange of specimens
  • Limited Lifetime Warranty
SPECS & DESCRIPTION
Specifications & Description
  • Magnification4x/10x/40x/100x
  • Maximum Magnification1000x
  • Head ConfigurationBinocular
  • ObjectivesSemi-plan
  • Eyepiece Diameter (mm)23.2
  • Eyepiece Magnification10x
  • Stage Dimensions171 x 115 mm
  • Illuminator(s)30 W halogen
  • Power (VAC)100 to 240
  • DescriptionEconomical Compound Microscope; 4x/10x/40x/1000x; 110-240 V
  • WarrantyLTD Life Time
Limited Lifetime Warranty
Key Features
  • Smooth-operating, ball-bearing quintuple nosepiece provides effortless objective changes
  • Wide ceramic-coated stage for smooth exchange of specimens
  • Limited Lifetime Warranty
More About this Item
Economical Compound Microscopes
These economical microscopes with S. Plan optics are ideal for education and research laboratory applications. Feature 30 W halogen illumination and comfortable siedentopf viewing heads with 23.2-mm eyetubes, and F.N. 20, KHW10X widefield eyepieces.
What’s included: Abbe 1.25 N.A. condenser in dovetail mount, 4x/10x/40x/100x oil objectives, S. Plan objectives, auto-voltage sensing power supply, 6-ft detachable power cord, green filter, dust cover, and immersion oil.