Meiji Techno Microscope Eyepieces, Super Widefield, 10X, 23 mm Field of View; Pair
Representative image only.
Meiji Techno Mfr # MA502 – Item # WW-48402-58
Interchangeable eyepieces enhance flexibility of microscopes
  • Compatible with Meiji Techno EMZ, EMT, and EMF model microscopes
  • Limited Lifetime Warranty
PRODUCT OPTIONS
₹41,400.00INR / Pair of 1
SPECS & DESCRIPTION
Specifications & Description
  • Eyepiece Magnification10x
  • Field of View23 mm
  • Outside Diameter (cm)3.05
  • DescriptionMicroscope Eyepieces, Super Widefield, 10X, 23 mm Field of View; Pair
  • WarrantyLTD Life Time
Limited Lifetime Warranty
Key Features
  • Compatible with Meiji Techno EMZ, EMT, and EMF model microscopes
  • Limited Lifetime Warranty
More About this Item
These paired interchangeable widefield eyepieces provide added customization to the Meiji Techno EMZ, EMT, and EMF model microscopes. Easily change eyepiece magnification for a wide range of applications. Microscopes sold separately.
Eyepiece with 25 mm reticle mount is designed for designed for Meiji Techno EMZ, EMT, and EMF model microscopes.