Meiji Techno EMZ-1 Stereozoom microscope bodies; objectives, 1x to 3x
Representative image only.
Meiji Techno Mfr # EMZ-1 – Item # WW-48402-10
  • Limited Lifetime Warranty
SPECS & DESCRIPTION
Specifications & Description
  • Magnification1x to 3x
  • Maximum Magnification3x
  • Head ConfigurationBinocular
  • Objectives1x to 3x
  • DescriptionStereozoom microscope bodies; objectives, 1x to 3x
  • WarrantyLTD Life Time
Limited Lifetime Warranty
Key Features
  • Limited Lifetime Warranty
More About this Item
Get crisp, high-resolution images with excellent stereoscopic viewing. The 45° inclined head has adjustable diopter and interpupillary controls for easy observing. Rotate body a full 360° and lock into any position. The eyepiece diameters on all models are 25 mm ID x 34 mm OD.
Model 48402-10 is the most economical stereozoom unit and perfect for industrial and educational applications where continuous magnification is necessary.