Meiji Techno EMT-2 Dual-power microscope bodies; objectives, 1x/3x
Representative image only.
Meiji Techno Mfr # EMT-2 – Item # WW-48403-30
  • Limited Lifetime Warranty
PRODUCT OPTIONS
₹157,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Magnification1x/3x
  • Maximum Magnification3x
  • Head ConfigurationBinocular
  • Objectives1x/3x
  • DescriptionDual-power microscope bodies; objectives, 1x/3x
  • WarrantyLTD Life Time
Limited Lifetime Warranty
Key Features
  • Limited Lifetime Warranty
More About this Item
Get crisp, high-resolution images with excellent stereoscopic viewing. The 45° inclined head has adjustable diopter and interpupillary controls for easy observing. Rotate body a full 360° and lock into any position. The eyepiece diameters on all models are 25 mm ID x 34 mm OD.
Dual-power models feature paired objectives mounted in a revolving turret for easy switching between low and high magnification.