Meiji Techno C-mount adapter for Olympus 35-mm camera
Representative image only.
Meiji Techno Mfr # MA150/50+MA512+T2-6 – Item # WW-48402-84
Easily connect your Meiji Techno microscope to a camera
  • Compatible with Meiji Techno microscopes
  • Limited Lifetime Warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionC-mount adapter for Olympus 35-mm camera
  • WarrantyLimited Lifetime
Limited Lifetime Warranty
Key Features
  • Compatible with Meiji Techno microscopes
  • Limited Lifetime Warranty
More About this Item
Use C-mount adapters to attach a digital camera or video camera to the photo tube of your Meiji Techno microscope. The C-mount adapter that is needed will depend on your microscope, as well as your camera. Camera and microscope sold separately.