Meiji Techno Replacement Microscope Lamp, Mercury, HBO, 20V, 100W
Representative image only.
Meiji Techno Mfr # BA005 – Item # WW-48924-66
Optimize life and performance of your Meiji Techno microscopes
  • Easily replacement microscope bulb to keep your microscope working
  • Ideal replacement lamp for the epi-fluorescent microscope
  • Provides up to 200 hours of useful life
  • Limited Lifetime Warranty
PRODUCT OPTIONS
₹46,500.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Illuminator(s)Mercury
  • Power20V, 100W
  • DescriptionReplacement Microscope Lamp, Mercury, HBO, 20V, 100W
  • WarrantyLTD Life Time
Conformite EuropeenneLimited Lifetime Warranty
Key Features
  • Easily replacement microscope bulb to keep your microscope working
  • Ideal replacement lamp for the epi-fluorescent microscope
  • Provides up to 200 hours of useful life
  • Limited Lifetime Warranty
More About this Item
Keep your Meiji Techno microscopes running smoothly with these replacement bulbs.
The 20 volt, 100 watt mercury lamp features a SFa7.5-2 base.