Masterflex Valve, PVDF with EPDM Diaphragm, 4.0 Cv
Representative image only.
Masterflex Item # WW-40619-88
Fully drainable and easy to maintain!
  • Eliminate leaks—ideal for food or pharmaceutical processing
  • Compatible with Tri-Clamp® connections for easy maintenance, change out, and orientation—no cutting or welding needed!
  • Choose either natural polypropylene (PP) or Kynar® (PVDF) body—both are excellent for ultrapure applications
  • Models with Kynar® (PVDF) body offer additional chemical resistance to corrosives
PRODUCT OPTIONS
₹199,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Clamp Size (in)0.75
  • Outlet Port3/4" Tri-clamp
  • Connection TypeTri-clamp
  • Inlet Port3/4" Tri-clamp
  • Cv4.0
  • Max Pressure (PSI)125
  • Max Temperature (° F)210
  • Max Temperature (° C)99
  • Wetted MaterialsPVDF, EPDM
  • DescriptionValve, PVDF with EPDM Diaphragm, 4.0 Cv
Key Features
  • Eliminate leaks—ideal for food or pharmaceutical processing
  • Compatible with Tri-Clamp® connections for easy maintenance, change out, and orientation—no cutting or welding needed!
  • Choose either natural polypropylene (PP) or Kynar® (PVDF) body—both are excellent for ultrapure applications
  • Models with Kynar® (PVDF) body offer additional chemical resistance to corrosives
More About this Item
These high-purity valves feature manual actuators. Maximum viscosity is 400 cSt. Maximum fluid temperature is 180°F (82°C).