Masterflex Sanitary Gasket, EPDM, 1-1/2" Tri-Clamp®; 10/PK
Representative image only.
Masterflex Item # WW-30548-64
Ensure a tight seal
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Single-use custom configuration solutions to minimize downtime
  • Always in stock—most materials and configurations available to ship same day
  • Material: EPDM
  • Clamp Size: 1.5"
PRODUCT OPTIONS
₹3,800.00INR / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialEPDM
  • Connection TypeSanitary
  • Sanitary Gasket Size (in)1 1/2
  • DescriptionEPDM Sanitary Gasket, 1-1/2" Tri-Clamp; 10/Pack
Certificate of Compliance
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Single-use custom configuration solutions to minimize downtime
  • Always in stock—most materials and configurations available to ship same day
  • Material: EPDM
  • Clamp Size: 1.5"
More About this Item
These gaskets are for Tri-Clamp® style fittings and come in a wide variety of materials to match your specific process needs. All gaskets meet USDA, 3-A and CGMP criteria. FDA Compliant. All silicone, EPDM, PTFE, and Viton® gaskets are animal-derived ingredient free and meet USP Class IV and cytotoxicity criteria. Contact Cole-Parmer for other sizes or materials.
Platinum-cured silicone is ideal for most pharmaceutical, food, and beverage processing applications. Odorless and tasteless to ensure media purity. May undergo SIP. Usable from -58 to 446°F (-50 to 230°C).
Viton offers excellent chemical resistance, especially to many acids and alkalis.